Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

Tau (Total) Human Singleplex Kit

Catalog number: LHB0041

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of human Total Tau in CSF and tissue culture supernatant. The assay may be run alone or in combination with other Intracellular Singleplex Bead Kits. These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LNB0001). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: See product documentation
Assay Sensitivity: <22 pg/mL
Bead Region: 69
Former Identifiers: Novex
Gene Name: microtubule associated protein tau
Gene Symbol: MAPT
Immunoassay Kit Format: Singleplex Kit
Protein Aliases: Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau
Protein Name: Microtubule-associated protein tau
Target (Kit Named): Tau
Target Species (Named): Human
Target Species (Validated): Human
Target Sub-Specificity: Total
UniProt ID: P10636
Detection Method: Fluorescent
For Use With (Equipment): Luminex™ Instruments
Gene Aliases: DDPAC, FTDP-17L, MSTD, MTBT1, MTBT2, PPND, PPP1R103, TAU, MAPT
Gene ID: 4137
Label or Dye: R-PE
Product Size: 100 tests
Sample Type (Specific): Cell Culture Supernatants, Cerebrospinal Fluid
System: Luminex™
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products