Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

Amyloid beta 40 Human Kit for Luminex™ Platform

Catalog number: LHB3481

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of human Aβ40 (Beta Amyloid) in CSF and tissue culture media.The assay may be run alone or in combination with other Neuroscience Singleplex Bead Kits, but cannot be multiplexed with the Aβ42 Singleplex Bead Kit (Cat. No. LHB3441) or Aβ Aggregate Singleplex Bead Kit (Cat. No. LHB3491). These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LNB0001). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Marker: Aβ 40
System: Luminex®
Gene ID: 351
Species: Human
Bead Type: Polystyrene
Bead Region: 60
Gene Symbol: APP
Sensitivity: 22 pg/ml
Label or Dye: R-PE
Product Size: 100 tests
Detection Method: Fluorescent
Research Category: Neurobiology
Sample Type (Specific): Cell Culture Supernatants, Cerebrospinal Fluid
For Use With (Equipment): Luminex® Instruments
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products