This product will be discontinued once remaining inventory is depleted. We offer the IL-8 Human Ultrasensitive Magnetic Bead Kit as a replacement.

IL-8 Human Singleplex Kit, Ultrasensitive

Catalog number: LHC0083

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This antibody bead kit comprises the analyte specific components to detect and quantify protein levels of Human IL-8. IL-8 (Interleukin-8) is chemotactic for migratory immune cells and inhibits the adhesion of leukocytes to activated endothelial cells. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0003). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: See product documentation
Assay Sensitivity: <1 pg/mL
Bead Region: 81
Former Identifiers: Novex
Gene Name: C-X-C motif chemokine ligand 8
Gene Symbol: CXCL8
Immunoassay Kit Format: Singleplex Kit
Immunoassay Kit Grade: Ultrasensitive
Protein Name: Interleukin-8
Target (Kit Named): IL-8
Target Species (Named): Human
Target Species (Validated): Human
UniProt ID: P10145
Detection Method: Fluorescent
For Use With (Equipment): Luminex™ Instruments
Gene Aliases: GCP-1, GCP1, IL8, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1
Gene ID: 3576
Label or Dye: R-PE
Product Size: 100 tests
Sample Type (Specific): Serum
System: Luminex™
Target Protein: IL-8
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products