This product will be discontinued once remaining inventory is depleted. We offer the TNF alpha Human Magnetic Kit for Luminex Platform as a replacement.

TNF alpha Human Singleplex Kit

Catalog number: LHC3011

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This singleplex bead kit is specifically designed for the measurement of human TNF-α in serum, plasma, and tissue culture supernatant. This assay may be run alone or in combination with other select Luminex™ singleplex kits. The panel is suitable for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0001).
Superior performance—accurate, reproducible, and sensitive quantitation of multiple proteins
High quality—fully qualified antibodies permit excellent specificity and sensitivity
Fast and easy protocols—perform your assay and analyze your data typically in less than one day

Luminex™ xMAP™ Technology—Effective and Proven Analytical Tool
The TNF-α Human Singleplex Bead Kit is based on xMAP™ technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex™ assays. Polystyrene microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead is given a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format. Upon completion of the sandwich immunoassay, beads must be measured using a Luminex™ detection system (100™/200™ or FLEXMAP 3D™). The instrument uses xPONENT™ software to distinguish bead color (analyte) and R-PE fluorescence intensity (assay signal strength) to quantify target(s).

More Validation for Confidence in Your Results
The TNF-α Human Singleplex Bead Kit has been validated using the same rigorous criterion that distinguishes Luminex™ assays from other commercially available assays. With more than 20 years of experience, our fully qualified antibodies help ensure the specificity and sensitivity of our assays. When benchmarked against the competition, our multiplex products consistently deliver excellent results. Every singleplex kit and premixed multiplex kit comes with a product insert that outlines the specifications for each marker in the assay (see Documents section below). Standard curves (Figures 1 and 2) are generated for each lot of product to assure quantitation consistency.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: See product documentation
Assay Sensitivity: <10 pg/mL
Bead Region: 50
Former Identifiers: Novex
Gene Name: tumor necrosis factor
Gene Symbol: TNF
Immunoassay Kit Format: Singleplex Kit
Protein Name: tumor necrosis factor
Target (Kit Named): TNF alpha
Target Species (Named): Human
Target Species (Validated): Human
UniProt ID: P01375
Detection Method: Fluorescent
For Use With (Equipment): Luminex™ Instruments
Gene Aliases: DIF-alpha, TNFA, TNFSF2, TNLG1F, TNF
Gene ID: 7124
Label or Dye: R-PE
Product Size: 100 tests
Sample Type (Specific): Cell Culture Supernatants, Plasma, Serum
System: Luminex™
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products