Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

IFN alpha Human Kit for Luminex™ Platform

Catalog number: LHC4011

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of human IFN-&alpha (Interferon-alpha) in plasma or tissue culture supernatant. The assay may be run alone or in combination with other Human Singleplex Bead Kits. These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0001). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Marker: IFN-α
System: Luminex®
Gene ID: 3438
Species: Human
Bead Type: Polystyrene
Bead Region: 17
Gene Symbol: IFN1@
Sensitivity: <15 pg/ml
Target Gene: interferon, type 1, cluster
Gene Aliases: IFNA
Label or Dye: R-PE
Product Size: 100 tests
Protein Family: Cytokines & Receptors
Detection Method: Fluorescent
Research Category: Immunology, Signal Transduction
Sample Type (Specific): Plasma, Cell Culture Supernatants
For Use With (Equipment): Luminex® Instruments
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products