This product will be discontinued once remaining inventory is depleted. We offer the Cytokine Human Magnetic 30-Plex Panel as a replacement.

Cytokine 30-Plex Human Panel, Polystyrene

Catalog number: LHC6003

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Cytokine Human 30-Plex Panel for the Luminex™ platform is specifically designed for quantifying human cytokines, chemokines, and growth factors in serum, plasma, and tissue culture supernatant. By measuring 30 analytes simultaneously, the Luminex™ assay panel helps provide more data from each sample, saving both money and time compared to more traditional systems (such as ELISA) used in research. This assay may be run alone or in combination with other select Luminex™ singleplex kits. The panel is suitable for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Advantages of the The Cytokine Human 30-Plex Panel:

Superior performance—accurate, reproducible, and sensitive quantitation of multiple proteins
High quality—fully qualified antibodies permit excellent specificity and sensitivity
Fast and easy protocols—perform your multiplex assay and analyze your data typically in less than one day

Comprehensive human analyte panel
The Human Cytokine 30-Plex Panel for the Luminex™ platform provides reagents for the accurate, reproducible, and sensitive quantitation of human proteins including:

CytokinesChemokinesGrowth Factors
G-CSFEotaxinEGF
GM-CSFIP-10FGF-basic
IFN-αMCP-1HGF
IFN-γMIGVEGF
IL-1βMIP-1α
IL-1RAMIP-1β
IL-2RANTES
IL-2R
IL-4
IL-5
IL-6
IL-7
IL-8
IL-10
IL-12 (p40/p70)
IL-13
IL-15
IL-17
TNF-α

Luminex™ xMAP™ technology—effective and proven analytical tool
The Human Cytokine 30-Plex Panel is based on xMAP™ technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex™ assays. Polystyrene microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead is given a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format. Upon completion of the sandwich immunoassay, beads must be measured using a Luminex™ detection system (100™/200™ or FLEXMAP 3D™). The instrument uses xPONENT™ software to distinguish bead color (analyte) and R-PE fluorescence intensity (assay signal strength) to quantify target(s).

More validation for confidence in your results
The Human Cytokine 30-Plex Panel has been validated using the same rigorous criterion that distinguishes Luminex™ assays from other commercially available assays. With more than 20 years of experience, our fully qualified antibodies help ensure the specificity and sensitivity of our assays. When benchmarked against the competition, our multiplex products consistently deliver excellent results. Every singleplex kit and premixed multiplex kit comes with a product insert that outlines the specifications for each marker in the assay (see Documents section). Standard curves (see figures) are generated for each lot of product to assure quantitation consistency.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Former Identifiers: Novex
Detection Method: Fluorescent
For Use With (Equipment): Luminex™ Instruments
Immunoassay Kit Format: Panel
Immunoassay Kit Grade: Polystyrene
Label or Dye: R-PE
Multiplex Panel Name: Cytokine
Multiplex Size (Plex No.): 30
Product Size: 100 tests
Sample Type (Specific): Cell Culture Supernatants, Plasma, Serum
System: Luminex™
Target Species (Named): Human
Target Species (Validated): Human
Shipping Condition: Wet Ice

Contents & storage

Contains premixed antibody coated capture beads, standard, premixed detector antibody, diluents, SAV concentrate, buffers, wash solution, filter plate, complete protocol and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products