Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

EGF Human Kit for Luminex™ Platform

Catalog number: LHG0061

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of human EGF (Epidermal Growth Factor) in serum, plasma or tissue culture supernatant. The assay may be run alone or in combination with other Human Singleplex Bead Kits. These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0001). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Marker: EGF
System: Luminex®
Gene ID: 1950
Species: Human
Bead Type: Polystyrene
Bead Region: 8
Gene Symbol: EGF
Sensitivity: <15 pg/ml
Target Gene: epidermal growth factor
Gene Aliases: HOMG4, URG
Label or Dye: R-PE
Product Size: 100 tests
Protein Family: Growth Factors & Receptors
Accession Number: NM_001178130.1, NP_001171602.1, NM_001963.4, NM_001178131.1, NP_001171601.1, NP_001954.2
Detection Method: Fluorescent
Research Category: Immunology
Sample Type (Specific): Plasma, Cell Culture Supernatants, Serum
For Use With (Equipment): Luminex® Instruments
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products