ERK 1/2 [pT185/pY187] Human Kit for Luminex™ Platform

Catalog number: LHO0241

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of ERK1⁄2 (Extracellular signal-Regulated protein Kinase) when phosphorylated at threonine 185 and tyrosine 187 in cell lysates or tissue homogenates. This singleplex bead kit may be multiplexed with other phospho-specific and total protein singleplex bead kits available, but cannot be multiplexed with the ERK1⁄2 (Total) Singleplex Bead Kit (Cat. No. LHO0231). These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0002). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Marker: ERK1⁄2 [pTpY185⁄187]
System: Luminex®
Gene ID: 5595
Species: Human
Bead Type: Polystyrene
Bead Region: 57
Gene Symbol: MAPK3
Sensitivity: 0.5 U/ml
Target Gene: mitogen-activated protein kinase 3
Gene Aliases: ERK-1, ERK1, ERT2, HS44KDAP, HUMKER1A, MGC20180, PRKM3, p44-ERK1, p44-MAPK
Label or Dye: R-PE
Product Size: 100 tests
Protein Family: Kinases & Inhibitors
Protein Subtype: Serine⁄Threonine Kinases
Accession Number: NM_002746.2, NP_002737.2, NP_001035145.1, NM_001040056.1, NM_001109891.1, NP_001103361.1
Detection Method: Fluorescent
Research Category: Signal Transduction
Sample Type (Specific): Cell Extracts, Tissue Homogenates
For Use With (Equipment): Luminex® Instruments
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols