Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

GSK-3 beta (total) Multispecies Kit for Luminex™ Platform

Catalog number: LHO0451

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of total GSK-3β (Glycogen Synthase Kinase) in cell lysates and tissue homogenates. This singleplex bead kit may be multiplexed with other phospho-specific and total protein singleplex bead kits available, but cannot be multiplexed with the GSK-3β [pS9] Singleplex Bead Kit (Cat. No. LHO0461). These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0002). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Marker: GSK-3β (total)
System: Luminex®
Gene ID: 2932, 56637, 84027
Species: Mouse, Rat, Human
Bead Type: Polystyrene
Bead Region: 59
Gene Symbol: GSK3B, Gsk3b
Sensitivity: 0.18 ng/ml
Target Gene: glycogen synthase kinase 3 beta
Gene Aliases: 7330414F15Rik, 8430431H08Rik, C86142, GSK-3, GSK-3beta, GSK3
Label or Dye: R-PE
Product Size: 100 tests
Protein Family: Kinases & Inhibitors
Protein Subtype: Serine⁄Threonine Kinases
Detection Method: Fluorescent
Research Category: Signal Transduction
Sample Type (Specific): Cell Extracts, Tissue Homogenates
For Use With (Equipment): Luminex® Instruments
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products