Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

GSK3 beta (Phospho) [pS9] Multispecies Singleplex Kit

Catalog number: LHO0461

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of GSK-3β (Glycogen Synthase Kinase) when phosphorylated at serine 9 in cell lysates and tissue homogenates. This singleplex bead kit may be multiplexed with other phospho-specific and total protein singleplex bead kits available, but cannot be multiplexed with the total GSK-3β Singleplex Bead Kit (Cat. No. LHO0451). These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0002). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: See product documentation
Assay Sensitivity: 1.0 U/mL
Bead Region: 59
Former Identifiers: Novex
Gene Name: glycogen synthase kinase 3 beta
Gene Symbol: GSK3B
Immunoassay Kit Format: Singleplex Kit
Protein Aliases: GSK-3 beta, Serine/threonine-protein kinase GSK3B
Protein Name: Glycogen synthase kinase-3 beta
Target (Kit Named): GSK3 beta
Target Site-Specificity: [pS9]
Target Species (Named): Multispecies
Target Species (Validated): Human, Mouse, Rat
Target Sub-Specificity: Phospho
UniProt ID: P49841
Detection Method: Fluorescent
For Use With (Equipment): Luminex™ Instruments
Gene ID: 2932
Label or Dye: R-PE
Product Size: 100 tests
Protein Subtype: Serine⁄Threonine Kinases
Sample Type (Specific): Cell Extracts, Tissue Homogenates
System: Luminex™
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols