Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

IGF-1R [pY1135/pY1136] Human Kit for Luminex™ Platform

Catalog number: LHO0501

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of IGF-1R (Insulin-like Growth Factor 1 Receptor) when phosphorylated at tyrosine 1135 and tyrosine 1136 in cell lysates and tissue homogenates. This singleplex bead kit may be multiplexed with other phospho-specific and total protein singleplex bead kits available, but cannot be multiplexed with the total IGF-1R Singleplex Bead Kit (Cat. No. LHO0491). These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0002). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Marker: IGF-1R [pYpY1135⁄1136]
System: Luminex®
Gene ID: 3480, 16001, 25718
Species: Human
Bead Type: Polystyrene
Bead Region: 73
Gene Symbol: IGF1R, Igf1r
Sensitivity: 1.3 U/ml
Target Gene: insulin-like growth factor 1 receptor, insulin-like growth factor I receptor
Gene Aliases: A330103N21Rik, CD221, D930020L01, IGF-1R, hyft, IGFIRC, JTK13, CD221, IGFIR, IGFR, JTK13, MGC142170, MGC142172, MGC18216
Label or Dye: R-PE
Product Size: 100 tests
Protein Subtype: Receptor Tyrosine Kinases
Accession Number: NM_010513.2, NM_000875.3, NM_052807.2, NP_434694.1, NP_000866.1, NM_152452.1, NP_034643.2
Detection Method: Fluorescent
Research Category: Signal Transduction
Sample Type (Specific): Cell Extracts, Tissue Homogenates
For Use With (Equipment): Luminex® Instruments
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products