Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

IR [pY1162/pY1163] Multispecies Kit for Luminex™ Platform

Catalog number: LHR9131

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of IR (Insulin Receptor) when phosphorylated at tyrosine 1162 and tyrosine 1163 in cell lysates and tissue homogenates. This singleplex bead kit may be multiplexed with other phospho-specific and total protein singleplex bead kits available, but cannot be multiplexed with the total IR Singleplex Bead Kit (Cat. No. LHR9111). These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0002). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Marker: IR [pYpY1162⁄1163]
System: Luminex®
Gene ID: 3643
Species: Mouse, Human, Rat
Bead Type: Polystyrene
Bead Region: 72
Gene Symbol: INSR
Sensitivity: 0.8 U/ml
Target Gene: insulin receptor
Gene Aliases: CD220, HHF5
Label or Dye: R-PE
Product Size: 100 tests
Protein Subtype: Receptor Tyrosine Kinases
Accession Number: NM_000208.2, NP_000199.2, NM_001079817.1, NP_001073285.1
Detection Method: Fluorescent
Research Category: Signal Transduction
Sample Type (Specific): Cell Extracts, Tissue Homogenates
For Use With (Equipment): Luminex® Instruments
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products