Human Neuroscience Buffer Reagent Kit

Catalog number:  LNB0001

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Buffer Reagent Kit comprises of the reagents that are intended for use with Invitrogen Human Neuroscience Singleplex Bead Kits for in vitro quantitative determination of analytes in human cerebrospinal fluid (CSF) and tissue culture medium. Invitrogen Singleplex Bead Kits have been developed to maximize flexibility in experimental design, permitting the measurement of one or multiple analytes in panels designed by the researcher. These reagents are intended for use in the Luminex® 100™ or 200™ System only.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
For Use With (Equipment): Luminex® Instruments
Product Line: Luminex™
Product Size: 100 tests
Readout Enzyme: RPE
Research Category: Neurobiology
Sample Type (Specific): Cell Culture Supernatants, Cerebrospinal Fluid
Species: Human
System: Luminex®
Shipping Condition: Wet Ice

Contents & storage

Contains diluents, SAV concentrate, buffers, wash solution, filter plate and complete protocol. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products