Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Marina Blue® 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Marina Blue® DHPE)

Catalog number: M12652

Invitrogen™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The phospholipid, Marina Blue® DHPE is labeled on the head group with the blue-fluorescent Marina Blue® fluorophore with excitation/emission maxima ~365/460 nm.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation⁄Emission (nm): 365⁄460
Product Line: MARINA BLUE®
Product Size: 1 mg
Reagent Type: Phospholipids
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols