Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Mag-Fura-2, AM, cell permeant

Catalog number:  M1291

Invitrogen™  Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Mag-fura-2, AM is an intracellular magnesium indicator that is ratiometric and UV light-excitable. This acetoxymethyl (AM) ester form is useful for noninvasive intracellular loading and is also available in special packaging (M1292). A Cell-impermeant tetrapotassium (M1290) salt form of the dye is available.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Calcium Indicator, Magnesium Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Flow Cytometer, Fluorescence Microscope, Microphotometer
Cell Permeability: Cell-Permeant
Detection Method: Fluorescent
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols