Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Mag-Fluo-4, AM, cell permeant - Special Packaging

Catalog number: M14206

Molecular Probes™  Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The cell-permeant mag-fluo-4, AM is an analog of fluo-4 with a Kd for Mg2+ of 4.7 mM and a Kd for Ca2+ of 22 µM, making it useful as an intracellular Mg2+ indicator as well as a low-affinity Ca2+ indicator.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Calcium Indicator, Magnesium Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope, Microphotometer, Flow Cytometer
Cell Permeability: Cell-Permeant
Product Size: 10 x 50 µg
Detection Method: Fluorescent
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols