Multiplexed Proteomics™ Phosphoprotein Gel Stain Kit #2 (with Pro-Q™ Diamond and SYPRO™ Ruby Gel stains)

Catalog number:  M33306

Invitrogen™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Our new Pro-Q Diamond phosphoprotein gel stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein specific antibodies and Western blot analysis. This reagent, when combined with our total protein stain – SYPRO Ruby protein gel stain – becomes a powerful combination for quantitative proteome analysis. This convenient Pro-Q Diamond phosphoprotein gel stain and SYPRO Ruby protein gel stain pack provides each stain in 200 mL amounts. This combination is also available in 1 liter amounts (M-33305).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Target Molecule: Proteins (Phosphoproteins)
Label or Dye: Pro-Q® Diamond, SYPRO® Ruby
Detection Method: Fluorescent
Detection Location: In-Gel Detection
Product Line: PRO-Q®, SYPRO®
Product Size: 1 set
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products