MitoProbe™ DiIC1(5) Assay Kit - For Flow Cytometry - 100 Assays

Catalog number: M34151

Invitrogen™  Related applications: Cell Structure | Cell Viability, Proliferation & Function | Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The MitoProbe™ DiIC1(5) Assay Kit provides solutions of the membrane-potential-sensitive cyanine dye DiIC1(5), and a mitochondrial membrane-potential disrupter, CCCP, for the study of mitochondrial membrane potential. DiIC1(5) penetrates the cytosol of eukaryotic cells and accumulates primarily in mitochondria with active membrane potentials, producing bright, far-red fluorescence. DiIC1(5) staining intensity decreases with decreased mitochondrial membrane potential.

View a selection guide for all apoptosis assays for flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: DiIC1(5)
Sub-Cellular Localization: Mitochondria
Detection Method: Fluorescent
Excitation⁄Emission (nm): 638⁄658
Flow Cytometer Laser Lines: 633/635
For Use With (Equipment): Flow Cytometer
Number of Reactions: 100
Product Line: MitoProbe™
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Contains 1 vial of DiIC1(5) ( 625 µL, 10 µM in DMSO), and 1 vial of CCCP: (125 µL, 50 mM DMSO).

Store at 2–6°C, protected from light. Do not freeze.

Documents

Manuals & protocols

Recommended products