Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

MEQ (6-Methoxy-N-Ethylquinolinium Iodide)

Catalog number:  M6886

Invitrogen™  Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The chloride indicator MEQ has absorption/emission maxima ~344/440. MEQ can be reduced to create the cell-permeant dihydro-MEQ (DiH-MEQ) which can be passively loaded into cells and subsequently oxidized to the chloride-sensitive MEQ.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Chloride Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope, Microplate Reader
Cell Permeability: Cell-Permeant
Detection Method: Fluorescent
Product Size: 100 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols