MethylCode™ Bisulfite Conversion Kit

Catalog number: MECOV50

Applied Biosystems™  Related applications: Epigenetic Sequencing | Methylation Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Please see our newest bisulfite conversion kit, Cells-to-CpG™ Bisulfite Conversion System, designed to provide more flexibility in starting samples without the need to purify your DNA and more confidence with available methylation controls. Cells-to-CpG™ is completely validated for Life Technologies' downstream platforms and replaces MethylCode™ as the bisulfite conversion kit of choice.

The MethylCode™ Bisulfite Conversion Kit is a fast and easy way to convert unmethylated cytosines from a DNA sample into uracils while methylated cytosines remain unchanged.

The modified DNA is ideal for PCR amplification for downstream analyses, including restriction endonuclease digestion, sequencing, and microarrays. With the MethylCode™ Bisulfite Conversion Kit you can expect:

–Simple, rapid cytosine methylation detection

–Consistency across most experimental conditions
–Streamlined conversion of unmethylated cytosines into uracil, thanks to a coupled DNA denaturation/bisulfite treatment step
–Ultra-pure DNA ideal for subsequent molecular-based analyses



Kit specifications:

–Protocol: Rapid heat denaturation of DNA followed by sodium bisulfite modification with spin column purification

–Sample sources: Plasmid, genomic, and endonuclease digested DNAs
–Input DNA: 500 pg-2 µg DNA per treatment, with 200-500 ng being optimal
–Recovery volume: Purified, bisulfite-treated DNA is recovered in 10 µl elution buffer



Contents and storage:
The MethylCode™ Bisulfite Conversion Kit contains all the reagents necessary to convert your DNA for methylation studies. The kit includes CT Conversion Reagent, Dilution Buffer, Resuspension buffer, Binding Buffer, Wash Buffer, Desulphonation Buffer, Elution Buffer, Spin Columns, Collection Tubes.

Store at room temperature for up to 24 months.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: MethylCode™
Product Size: 50 reactions
Sample Type (Specific): DNA (Genomic)
Starting Material (Cell #): Purified genomic DNA - 200 ng to 500 ng
Shipping Condition: Room Temperature

Contents & storage

The MethylCode™ Bisulfite Conversion Kit contains all the reagents necessary to convert your DNA for methylation studies. The kit includes CT conversion reagent, dilution buffer, resuspension buffer, binding buffer, wash buffer, desulphonation buffer, elution buffer, spin columns, and collection tubes.

Documents

Manuals & protocols

Recommended products