EpiLife® CF Kit This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: MEPICF500

Gibco™  Related applications: Clinical & Translational Research | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A sterile, liquid medium prepared without calcium chloride for the long-term, serum-free culture of human epidermal keratinocytes and human corneal epithelial cells. This basal medium requires the addition of calcium plus an appropriate supplement prior to use. Calcium chloride is provided as a separate component with each bottle of medium. EpiLife greatly extends the in vitro lifespan of human epidermal keratinocytes compared to other serum-free media systems. This basal media is animal origin free and contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. It does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. EpiLife® medium is HEPES and bicarbonate buffered, and is designed for use in an incubator with an atmosphere of 5% CO2 and 95% air.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Species: Human
Cell Type: Keratinocytes, Epithelial Cells (Corneal)
Serum Level: Serum-Free
Product Size: 500 mL
Classification: Animal Origin-Free, Protein-Free, Chemically Defined, Serum-Free
Inorganic Salts: No Calcium
Phenol Red Indicator: Phenol Red
Green Features: Fewer resources used, Less waste

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Media formulations

Documents

Manuals & protocols

Recommended products