Invitrogen™

Mouse IgM, (FITC)

Catalog number:  MGM01

 Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Isotype: Mouse IgM
Class: Class I (ASR)
Statement: : ASR: Analyte Specific Reagent. Analytical and performance characteristics are not established.
Buffer: PBS
Preservative: 0.1% Sodium Azide
Analyte Specific Reagent. Analytical and performance characteristics are not established.

Specifications

Host Species: Mouse
SubClass (Isotype): IgM
Antibody Fragment: Whole Antibody
Label or Dye: FITC (Fluorescein)
Number of Reactions: 50 Tests
Product Size: 0.5 mL
Shipping Condition: Wet Ice
Validated Application: Flow Cytometry

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products