Thermo Scientific™
microPHAZIR™ PC Analyzer for Plastic/Polymer Identification
 Related applications: Spectroscopy Elemental Isotope Analysis
Recycling of plastics, carpeting and other polymer materials is increasing as more of these products are being recovered, recycled and reused by consumers and recycling facilities. Post-industrial and post-consumer materials need to be accurately identified and sorted to achieve high quality reprocessed material. The Thermo Scientific™ microPHAZIR™ PC Analyzer for Plastic/Polymer Identification is an NIR spectroscopy tool that rapidly screens and identifies a variety of polymers, plastics and fiber types on-site to streamline inspection without compromising accuracy and quality. 
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The 2.75 lb (1.25 kg) microPHAZIR PC Analyzer for Plastic/Polymer Identification is a completely self-contained, powerful and cost-effective tool for the proper sorting and recycling of post-industrial and post-consumer polymer materials and carpet fibers.
Save Time
Rapid and accurate results displayed within seconds.
Easy to Use
Simple point-and-shoot operation designed for non-expert users, the analyzer is fully automated, precalibrated, and requires no user input.
Portable
Small and lightweight for fast identification of materials in the field or at the sorting facility.
Safe
No sample preparation or burn test necessary, NIR is fast, safe and nondestructive.
Identification of a variety of plastic types, including: 
PLA, PET, PP, PS, ABS, PI, PSO, PE, PPS, TPV, PTT, PC, PMP, PBT, PA, PETG, SAN, EVA, PB, PPO, CA, Nylon, PMMA, PUR, PI, PVC, PVA, PLA, Ionomer, Pollyalomer, Vinyl, Styrenic terpolymer, Elastomer
Need product support?
Get software updates, find support resources and manage smart, connected products and services for portable analytical instruments.
Get support
NIR technology
Near Infrared (NIR) can be very useful in probing bulk material by penetrating much farther into a sample than mid infrared radiation with little or no sample preparation. 
Learn about NIR ›
Documents
Manuals & protocols