New Transfection Support Center—find technical tips and troubleshooting recommendations from our scientists.
Invitrogen™

Neon™ Transfection System 100 µL Kit

Catalog number:  MPK10025

 Related applications: RNAi | Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Neon® Transfection System 100 µL Kit is designed specifically for use with the Neon® Transfection System. Use this kit for transfection volumes of 100 µL, containing 5 × 105–2 × 106 adherent cells or 1 × 106–5 × 106 suspension cells. Cells that have been transfected using the included 100 µL Neon® Tips are then ready to be washed and plated into a 6-well, 60 mm, or 10 cm culture dish, depending on your protocol.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Established Cell Lines, Hard-to-Transfect Cells, Primary Cells, Stem Cells
Classification: Animal Origin-Free, Chemically Defined, Low-Protein, Reduced Growth Factors, Reduced Serum, Xeno-Free
Compatible Products: Transfection Grade Nucleic Acid purification kits, siRNA vectors, DNA vectors
For Use With (Equipment): Neon® Transfection System
Format: Electroporation Pipette Tip
Product Line: Neon®
Product Size: 25 x 2 reactions
Sample Loading Volume: 100µl
Sample Type (Specific): Plasmid DNA, Synthetic siRNA
Serum Compatible: Yes
Shipping Condition: Room Temperature
Starting Material (Cell #): 2x10^4 - 6 x10^8 cells; Varies by cell type
System: Neon® Transfection System
Transfection Technique: Electroporation

Contents & storage

The Neon® Transfection System 100 µL Kit comes with 10 mL resuspension buffer R, 10 mL resuspension buffer T, 75 mL E2 electrolytic buffer, 25 x 100µL reaction tips, 5 electroporation tubes. Refer to the Neon® Transfection System users manual for protocol instructions. Store at room temperature.

Documents

Manuals & protocols