Pro-Q® Diamond Phosphoprotein Gel Staining Kit (includes 200 mL Stain, 40 µL Standard)

Catalog number: MPP33301

Invitrogen™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Pro-Q® Diamond Phosphoprotein Gel Staining Kit, includes 200 mL of the Pro-Q® Diamond Phosphoprotein Gel Stain and 40 µL of the PeppermintStick™ phosphoprotein molecular weight standards. The Pro-Q® Diamond phosphoprotein gel stain is a fluorescent stain (excitation/emission maxima ~555/580 nm) that can detect as little as 1-16 ng of phosphoprotein per band with a signal that is linear over 3 orders of magnitude. The PeppermintSTick™ phosphoprotein molecular weight standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Target Molecule: Proteins (Phosphoproteins)
Label or Dye: Pro-Q® Diamond
Detection Method: Fluorescent
Detection Location: In-Gel Detection
Product Line: PRO-Q®
Product Size: 200 mL Stain, 40 µL Standard

Contents & storage

Contains 200 mL of the Pro-Q® Diamond Phosphoprotein Gel Stain and 40 µL of the PeppermintStick™ phosphoprotein molecular weight standard

Documents

Manuals & protocols

Recommended products