Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

NeuroTrace™ 530/615 Red Fluorescent Nissl Stain - Solution in DMSO

Catalog number:  N21482

 Related applications: Cell Structure | Cell Tracing & Tracking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Nissl staining is a standard histological method for visualizing neurons in the brain and spinal cord. Composed of ribosomal RNA associated with the rough endoplasmic reticulum in neuronal perikarya and dendrites, the Nissl substance redistributes within the cell body in injured or regenerating neurons, providing a marker for the physiological state of the neuron. Our NeuroTrace 530/615 red fluorescent Nissl stain is selective for the Nissl substance characteristic of neurons and provides more sensitivity than traditional histological dyes like toluidine blue or cresyl violet.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Orange
For Use With (Equipment): Fluorescence Microscope
Sub-Cellular Localization: Nissl Bodies
Detection Method: Fluorescent
Excitation⁄Emission (nm): 530⁄615
Product Line: NEUROTRACE®
Product Size: 1 mL
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products