Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
Invitrogen™

NeuroTrace™ DiO Tissue-Labeling Paste

Catalog number:  N22881

 Related applications: Cell Tracing & Tracking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NeuroTrace tissue labeling pastes consist of DiI (N-22880), DiO or DiD (N-22882) mixed into an inert, water-resistant gel. The pastes are ready to use as supplied and can be applied directly to live or fixed tissue specimens using the tip of a needle.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Green
For Use With (Equipment): Fluorescence Microscope
Sub-Cellular Localization: Cell Membranes & Lipids
Detection Method: Fluorescent
Product Line: NEUROTRACE®
Product Size: 500 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products