New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

NanoOrange™ Protein Quantitation Kit, 200-2,000 assays

Catalog number:  N6666

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The NanoOrange Protein Quantitation Kit contains a very sensitive and easy assay for protein quantitation, with detection as low as 10 ng/mL of protein in solution. This fluorescent dye is suitable for use with spectrofluorometers and microplate readers. For detection of lipoproteins or proteins in a complex lipid environment, check out our CBQCA Protein Quantitation Kit (C-6667).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: Protein Quantitation Assay
Detection Method: Fluorescent
For Use With (Equipment): Microplate Reader
Product Line: NanoOrange®
Product Size: 1 kit

Documents

Manuals & protocols

Recommended products