Get the sharpest, brightest images. Try new Invitrogen™ ProLong™ Glass antifade mountant with glass-matching refractive index of 1.52. Learn more
Invitrogen™

Nucleic Acid Stains Dimer Sampler Kit

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Nucleic Acid Stains Dimer Sampler Kit contains 25 µL each of the following high-affinity nucleic acid stains based upon symmetric dimers of cyanine dyes: BOBO-1 iodide (Cat. No. B3582), BOBO-3 iodide (B3586), POPO-1 iodide (P3580), POPO-3 (P3584), TOTO-1 iodide (T3600), TOTO-3 iodide (T3604), YOYO-1 iodide (Y3601), and YOYO-3 iodide (Y3603).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Blue, Green, Orange, Red, Yellow
For Use With (Equipment): Fluorescence Microscope
Cell Permeability: Cell-Impermeant
Sub-Cellular Localization: Nucleic Acids
Detection Method: Fluorescent
Excitation⁄Emission (nm): various
Label or Dye: BOBO™-1, BOBO™-3, POPO™-1, POPO™-3, TOTO®-1, TOTO®-3, YOYO®-1, YOYO®-3
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Contains 25 µL of a 1 mM solution in DMSO of each dye in separate vials.

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products