Rat Glial Precursor Cells

Catalog number: N7746100

Gibco™  Related applications: Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ready-to use system: Upon proliferation in StemPro® NSC-SFM, >80% of the cells retain their undifferentiated Phenotype, good post-thaw cell viability (≥ 50%), and supports differentiation into Oligodendrocytes and Astrocytes. Gibco® Rat Glial Precursor Cells (GPC) offer researchers ready access to high quality cells and eliminate the time and effort to isolate these highly sensitive cells in house. Glial Precursor Cells are isolated from the cortex of newborn Sprague-Dawley rats. The cells are isolated under sterile conditions, expanded in complete StemPro® NSC SFM, and cryopreserved at passage 2 (P2). Each vial contains 1 × 106 cells/ml viable cells (post-thaw) that can be expanded once to give >2 fold increase of cells. Upon proliferation in StemPro® NSC SFM, >80% of cells retain their undifferentiated Phenotype and can be differentiated into Oligodendrocytes and Astrocytes. Glial Precursor Cells are the restricted progenitors, and the majority of their down stream progeny are Oligodendrocytes and Astrocytes. Oligodendrocytes produce myelin to protect neurons and secure proper signal transduction. Demyelination is involved with critical diseases, such as multiple sclerosis, Addison-Schilder Disease (ALD) etc.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Age: Newborn (14 days or less)
Species: Rat
Quantity: 1 each
Cell Type: Precursor Cells
Culture Type: Adherent Cell Culture
Product Size: 1 mL
Classification: Serum-Free
Number of Cells: 1 x 106
Donor Attributes: Normal
Cell Passage Number: Passage 2
Media Recommendation: StemPro® NSC SFM

Contents & storage

One vial containing ~1 x 10^6 cells; store in liquid nitrogen

Documents

Manuals & protocols

Recommended products