Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
Invitrogen™

Newport Green™ DCF, Dipotassium Salt, cell impermeant

Catalog number:  N7990

 Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Newport Green™ DCF indicator has moderate zinc-binding affinity (Kd for Zn2+ ~1 µM) but is essentially insensitive to Ca2+ (Kd for Ca2+ >100 µM), making this a valuable probe for detecting Zn2+ influx into neurons through voltage- or glutamate-gated channels. When used alongside dyes with dual Ca2+ / Zn2+ sensitivity such as fura-2 and mag-fura-2, Newport Green™ DCF provides confirmation that changes in Zn2+ levels, and not Ca2+ or Mg2+, are being detected. The response of Newport Green™ DCF to transition metals, including Zn, Mn, Fe, Co, Cu(I), Cu(II), Ni and Cd, has been characterized.

Newport Green™ PDX has a higher Zn2+ dissociation constant (Kd for Zn2+ ~30 µM) and a larger Zn2+-free to Zn2+-saturated fluorescence intensity increase than the Newport Green™ DCF indicators.

Fluorescent Zinc Indicators Specifications:
• Label (Ex/Em): Newport Green™ DCF (~505/535 nm)
• Kd (Zn2+) (in buffer):~1 µM
• Lyophilized product may be dissolved in distilled water or aqueous buffer for use
• Product is typically loaded into cells using microinjection or diffusion from patch pipette


Find Fluorescent Indicators for Zinc and Other Metal Ions
We offer a number of fluorescent indicators to determine polyvalent cation concentrations inside cells, to follow metal ion transport through ion channels, to make measurements in environmental samples, and more. Review Fluorescent Indicators for Zn2+ and Other Metal Ions—Section 19.7 in the Molecular Probes® Handbook for more information on these products.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Zinc Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope
Cell Permeability: Cell-Impermeant
Detection Method: Fluorescent
Product Line: Newport Green™
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products