MicroAmp® 96-Well Base

Catalog number: N8010531

Applied Biosystems™  Related applications: PCR | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® MicroAmp® Bases are designed to hold MicroAmp® Reaction Tubes in Tray/Retainer Sets or MicroAmp® 96-Well Reaction Plates for convenient use in pre- and post-PCR sample processing.

• Convenient for pre- and post-PCR sample processing
• Easy to use for pipetting steps
• Available in 24-well and 96-well plate formats, offering flexibility

Uniform Results
Use with all instruments that have heated sample covers, to eliminate condensation and the need for a mineral oil overlay. Proprietary MicroAmp® Reaction Tubes are essential for high-performance PCR. They ensure the greatest temperature and amplification uniformity for your samples.

Note: See user's manual or package insert for limited label license and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Black
For Use With (Equipment): 2720 Thermal Cycler, 7000 System, 7300 System, 7500 Fast Dx System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio™ Dx, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti® Dx Fast Thermal Cycler, Veriti® Dx Thermal Cycler, Veriti® Fast Thermal Cycler, Veriti® Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA™ 7 Dx System, ViiA™ 7 Fast System, ViiA™ 7 System
Plate Compatibility: 96-Well Plates
Product Line: MicroAmp®
Product Size: 10 pieces
Shipping Condition: Room Temperature
Type: Tube Tray

Contents & storage

Contains 10 MicroAmp® 96-Well Bases. Store at room temperature.

Documents

Manuals & protocols

Recommended products