Need help? Find instrument-specific tips, troubleshooting help, and technical resources within our new Real-Time PCR and Digital PCR Instruments Support Center.

MicroAmp™ Optical 96-Well Reaction Plate

Catalog number: N8010560

Applied Biosystems™  Related applications: PCR | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Applied Biosystems® MicroAmp® Optical 96-Well Reaction Plate is designed to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems® 96-well Real-Time PCR systems and thermal cyclers.

• Frosted to minimize interfering fluorescence from wells of cycling block
• Screened to eliminate auto-fluorescent plates
• Unique design provides a barrier to ambient air to help ensure well-to-well temperature uniformity

Oil-Free PCR Amplification
MicroAmp® disposables are optimized to provide unmatched temperature accuracy and uniformity for fast, oil-free PCR amplification. Proprietary MicroAmp® plates are essential for high-performance PCR and help ensure the greatest temperature and amplification uniformity for your samples.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Barcode: No Barcode
Capacity: 96 x 0.2 ml tubes
Color: Optical
For Use With (Equipment): 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 6100 Nucleic Acid PrepStation, 7000 System, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio™ Dx, SimpliAmp™ Thermal Cycler, Veriti® Dx Thermal Cycler, Veriti® Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System
Format: 96-well plate
Product Line: MicroAmp®
Product Size: 10 plates
Reaction Speed: Standard
Shipping Condition: Room Temperature

Contents & storage

Contains ten 96-well reaction plates.

Will require either caps or full plate cover to seal (not included).

May be stored at room temperature. With proper storage, plates may be stored indefinitely.

Documents

Manuals & protocols

Recommended products