GeneAmp® 10X PCR Buffer II & MgCl2

Catalog number: N8080010

Applied Biosystems™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® GeneAmp® 10X PCR Buffer II is optimized for use with AmpliTaq® DNA Polymerase, resulting in robust PCR amplification. The provided MgCl2 solution offers versatility for optimizing PCR.

• Optimized for use with AmpliTaq® DNA Polymerase, offering maximum PCR performance
• Available in several convenient sizes
• Separate MgCl2 solution offers versatility

GeneAmp® 10X PCR Buffer II, when diluted 1:10, provides the optimal pH and ionic strength for PCR amplification. GeneAmp® 10X PCR Buffer II is magnesium ion free. A separate MgCl2 solution is provided. When used together, GeneAmp® 10X PCR Buffer II and MgCl2 solution offer more versatility in optimizing the magnesium ion concentration to achieve optimal PCR amplification with any specific set of primers and DNA template. Note: See user's manual or package insert for limited label license and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: AmpliTaq® DNA Polymerase
Product Line: GeneAmp®
Product Size: 1 x 1.5 mL
Shipping Condition: Dry Ice

Contents & storage

1x 1.5 mL vial each of GeneAmp® 10X PCR Buffer II and MgCl2 sufficient for 100 PCR amplifications of 100µL each.

Store at -15 to -25°CC.

Documents

Manuals & protocols