GeneAmp® 10X PCR Buffer

Catalog number: N8080129

Applied Biosystems™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® GeneAmp® 10X PCR Buffer I is optimized for use with AmpliTaq® DNA Polymerase, resulting in robust PCR amplification.

• Optimized for use with AmpliTaq® DNA Polymerase, offering maximum PCR performance
• Available in several convenient sizes

GeneAmp® 10X PCR Buffer I contains 15 mM MgCl2. When diluted 1:10, it provides the optimal pH and ionic strength for PCR amplification with AmpliTaq® DNA Polymerase.

Note: See user's manual or package insert for limited label license and trademark information. For Research Use Only. Not for use in diagnostics procedures.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: AmpliTaq® DNA Polymerase
Product Line: GeneAmp®
Product Size: 6 x 1.5 mL
Shipping Condition: Dry Ice

Contents & storage

6 x 1.5 mL of GeneAmp® 10X PCR Buffer I (contains 15 mM MgCl2).

Store at -15 to -25°C.

Documents

Manuals & protocols

Recommended products