Applied Biosystems™

AmpliTaq™ DNA Polymerase with Buffer II (3000 units/tube)

Catalog number:  N8080153

 Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

AmpliTaq® DNA Polymerase is a 94 kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is the most thoroughly characterized enzyme available for PCR and its recombinant nature and purification method provide unparalleled purity and reproducibility, vial-to-vial, lot-to-lot.

Features of this enzyme:

  • AmpliTaq® DNA Polymerase is the most thoroughly characterized enzyme available for the PCR process, a testimony to its overall utility and efficacy
  • Its thermal activity profile makes it good for PCR applications
  • It is QC-tested to guarantee reproducible results

  • Reliable and robust PCR
    The thermal activity profile of AmpliTaq® DNA Polymerase is reliable for PCR applications because its optimal activity is in the same range at which stringent annealing of primers occurs (55–75°C). The enzyme's half-life is ~40 minutes at 95°C, providing thermostability that meets the requirements of most difficult PCR applications. This AmpliTaq® DNA Polymerase is supplied with GeneAmp® 10X PCR Buffer II. It is also available with GeneAmp® 10X PCR Buffer I and MgCl2 Solution.
    For Research Use Only. Not for use in diagnostic procedures.

    Specifications

    Polymerase: AmpliTaq® DNA Polymerase
    Hot Start: No
    Fidelity (vs. Taq): 1 X
    Reaction Format: Separate Components
    Product Overhang: 3'-A
    Reaction Speed: Standard
    Final Product Size(s): 5 kb or less
    Exonuclease Activity: 5' - 3'
    Product Line: AmpliTaq™
    Product Size: 3,000 units
    Shipping Condition: Dry Ice

    Contents & storage

    Contents:
    • 12 × 50 µL each of 5 U/µL of AmpliTaq® DNA Polymerase
    • 12 × 1.5 mL of GeneAmp® 10X PCR Buffer II (100 mM Tris-HCl pH 8.3, 500 mM KCl).
    • 12 × 1.5 mL 25 mM Magnesium Chloride

    Store at -20°C.

    Documents

    Manuals & protocols

    Recommended products