AmpliTaq™ DNA Polymerase, LD (Low DNA) with Buffer I

Numéro de catalogue:  N8080157

Applied Biosystems™  Applications liées: PCR

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

Applied Biosystems® AmpliTaq® DNA Polymerase, LD (Low DNA), is the same enzyme as AmpliTaq® DNA Polymerase, but is further purified through a proprietary separation process to ensure that residual bacterial DNA sequences are substantially reduced.

Manufacturing process results in highly purified enzyme with lower levels of bacterial DNA

• Minimizes non-specific PCR products when amplifying bacterial targets
• Highly purified enzyme making it particularly useful for low copy number amplifications

High-Quality
This highly purified enzyme preparation ensures that non-target, false-positive PCR products will be effectively minimized when amplifying bacterial sequences. This is especially useful for low-copy-number PCR amplifications. The enzyme is quality-control tested to verify that fewer than 10 copies of bacterial 16S ribosomal RNA gene sequences are present in a standard 2.5 unit aliquot. AmpliTaq® DNA Polymerase, LD is supplied with GeneAmp® 10X PCR Buffer I. It is also available with GeneAmp® 10X PCR Buffer II and MgCl2 Solution.

Note:
See user's manual or package insert for limited label license and trademark information. For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Polymerase: AmpliTaq® DNA Polymerase
Hot Start: No
Fidelity (vs. Taq): 1 X
Reaction Format: Separate Components
Product Overhang: 3'-A
Reaction Speed: Standard
Final Product Size(s): 5 kb or less
Exonuclease Activity: 5' - 3'
Product Line: AmpliTaq™
Product Size: 250 units
Purity or Quality Grade: LD (Low DNA)
Shipping Condition: Dry Ice

Contenu et stockage

Contains 250 units of AmpliTaq® DNA Polymerase, LD and Buffer I. Store at –15 to –30°C.

Documents

Manuels et protocoles

Produits recommandés