Large Gel Drying Kit
Catalog number:  NI2207
Invitrogen™  Related applications: Protein Gel Electrophoresis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Large Gel Drying Kit dries gels using the same principal components as the DryEase® Mini-Gel Drying System, but can accommodate larger format gels or four mini-gels. A base is not required for assembly.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line: Novex™
Product Size: 1 kit
Type: Gel Drying Accessories
Contents & storage
The Large Gel Drying Kit contains a DryEase® Large Gel Drying Frame (1 frame, 8 clamps), DryEase® Large Pre-Cut Cellophane (100 sheets), and Gel-Dry™ Drying Solution (500 ml). Store solution at room temperature. Guaranteed stable for one year when properly stored.
Documents
Manuals & protocols
Recommended products