Invitrogen™

DryEase™ Mini-Gel Drying Frame

Catalog number:  NI2380

 Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The DryEase® Mini-Gel Drying Frame is one component of the DryEase® Mini-Gel Drying System, which is a simple, easy-to-use kit for drying gels evenly using passive evaporation. The kit allows gels to be assembled on a base between two open frames, so that both outer surfaces of the gels are exposed to air. Consequently, drying is even and cracking is minimized. The proprietary Gel-Dry™ Drying Solution modulates the rate of water release to ensure crack-free gels. With the DryEase® System, gels are dried overnight with uniform tension on all points of the gel to eliminate cracking.

Applications: Gels dried with the DryEase® System are suitable for fluorography, densitometry, autoradiography, or permanent gel storage.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Novex™
Product Size: 2⁄pack
Type: Frame
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products