NuPAGE® Transfer Buffer (20X)

Catalog number: NP00061

Invitrogen™  Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NuPAGE® Transfer Buffer (20X) is a buffer used to transfer proteins from NuPAGE® Novex® gels to membranes for Western blotting. NuPAGE® Transfer Buffer can be used with the Novex® Semi-Dry Blotter for semi-dry transfer or with the XCell II™ Blot Module for wet (tank) transfer.

NuPAGE® Antioxidant may be used with NuPAGE® Transfer Buffer to enhance transfer of reduced proteins to membranes.

Related link
Learn more about other Invitrogen™ products for your protein transfer needs.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Type: NuPAGE® Gel
Shipping Condition: Room Temperature
Buffer Type: Transfer Buffers
Concentrated: 20 X
Gel Compatibility: NuPAGE® Gels
Product Line: NuPAGE®
Product Size: 1 L

Contents & storage

Store at room temperature. Guaranteed stable for 6 months unless otherwise specified in the product literature.

Documents

Manuals & protocols

Recommended products