New Protein Electrophoresis and Western Blotting Support Center—Find technical tips and troubleshooting recommendations from our scientists.
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

NuPAGE™ LDS Sample Buffer (4X)

Catalog number:  NP0007

 Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NuPAGE® LDS Sample Buffer (4X) is used to prepare protein samples for denaturing gel electrophoresis with NuPAGE® Novex® gels. Optimal protein sample preparation for polyacrylamide gel electrophoresis (PAGE) requires denaturing and reducing protein disulfide bonds. NuPAGE® LDS Sample Buffer contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.

Note: NuPAGE® LDS Sample Buffer should be brought to room temperature (25°C) prior to use. NuPAGE® LDS Sample Buffer is a highly viscous and concentrated solution—it contains twice the amount of LDS compared to the amount of SDS in Novex® Tris-Glycine SDS Sample Buffer or in Tricine SDS Sample Buffer. NuPAGE® LDS Sample Buffer also contains a higher concentration glycerol.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Type: NuPAGE Gel
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice
Buffer Type: Sample Loading Buffers
Concentrated: 4 X
Gel Compatibility: NuPAGE® Gels
Product Line: NuPAGE®
Product Size: 10 mL

Contents & storage

Contents: NuPAGE® LDS Sample Buffer (4X), 10 ml
Storage: +4°C to 25°C

Documents

Manuals & protocols

Recommended products