BioProbes® Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!

protein A, Alexa Fluor® 546 conjugate

Catalog number: P11049

Molecular Probes™  Related applications: Protein Assays and Analysis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Our Alexa Fluor 546 protein A conjugate binds to the Fc portion of various immunoglobulins from several different species. This exceptionally bright, orange-fluorescent probe (absorption/emission maxima ~556/573 nm) can be used to visualize antibodies in a manner analogous to a labeled species-specific secondary antibody.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor® 546
Product Line: Alexa Fluor®
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products