Larger pack sizes and volume discounts are available for this kinase protein. Inquire here.

PRKCB2 (PKC beta II) Recombinant Human Protein

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PKC family of serine⁄threonine kinases, including PRKCB2 (PKC beta II), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Kinases & Inhibitors
Research Category: Signal Transduction
Accession Number: NP_002729
Gene Aliases: PRKCB2 (PKC beta II)
Gene ID: 26191, 5579
Kinase Group: Ser⁄Thr Kinases: AGC Group
Molecular Weight: 77 kDa
Product Size: 5 µg
Protein Subtype: Serine⁄Threonine Kinases
Species: Human
System Compatibility: LanthaScreen® Antibody, LanthaScreen® Substrate, Omnia® Bulk Substrate, Z'-LYTE™
Target Gene: protein kinase C, beta 2
Validated Application: Kinase Assay
Protein Tag or Fusion: Untagged
Expression: Baculovirus
Kinase Family: PKC Kinase Family
Protein Form: Full Length, Recombinant
Shipping Condition: Dry Ice

Contents & storage

Storage Buffer: 50% glycerol, 20mM HEPES, pH 7.5, 2mM EDTA, 2mM EGTA, 5mM DTT, 250mM NaCl, 0.05% Triton X-100
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.

Documents

Manuals & protocols