PeppermintStick™ Phosphoprotein Molecular Weight Standards

Catalog number: P27167

Molecular Probes™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

PeppermintStick™ phosphoprotein molecular weight standards are a mixture of phosphorylated and nonphosphorylated proteins ranging from 14,400 to 116,250 molecular weight. Separation by polyacrylamide gel electrophoresis resolves the standard mixture into two phosphorylated and four nonphosphorylated protein bands. PeppermintStick™ phosphoprotein molecular weight standards are also available in a 400 µL volume (P33350}.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

System: SDS-PAGE
Size Range: 14.4 to 116.25 kDa
Stain Type: Unstained
Product Size: 40 µL
Ready to Load: No
Gel Compatibility: NuPAGE® Bis-Tris Gels, Novex® Tris-Glycine Gels, Novex® Tricine Gels, NuPAGE® Tris-Acetate Gels, SDS-PAGE Gels
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products