Kinase Quench Buffer

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Specifications

Form: Liquid
Product Size: 1 mL
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

Contents & storage

Kinase Quench Buffer (500 mM EDTA in water) may be is shipped frozen or at room temperature. Store at room temperature (20-30°C).

Documents

Manuals & protocols

Recommended products