Pro-Q® Diamond Phosphoprotein Gel Destaining Solution

Catalog number: P33310

Molecular Probes™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Pro-Q Diamond phosphoprotein gel stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein specific antibodies and Western blot analysis. Preparation of the Pro-Q Diamond phosphoprotein gel destaining solution is described in the gel staining protocol, but we also provide the detaining solution premixed for your convenience.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Target Molecule: Proteins (Phosphoproteins)
Label or Dye: Pro-Q® Diamond
Detection Method: Fluorescent
Detection Location: In-Gel Detection
Product Size: 1 L
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products