Invitrogen™
Pro-Q™ Diamond Phosphoprotein Enrichment & Detection Kit
Catalog number:  P33359
 Related applications: Protein Purification & Isolation
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Pro-Q® Diamond Phosphoprotein Enrichment and Detection Kit includes the Pro-Q® Diamond Phosphoprotein Enrichment Kit (P33356) for the isolation of phosphoproteins from complex cellular extracts, the Pro-Q® Diamond Phosphoprotein Gel Stain for detection of all O-linked phosphoproteins in polyacrylamide gels and the PeppermintStick™ phosphoprotein molecular weight standards (P27167).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line: PRO-Q®
Product Size: 1 kit
Documents
Manuals & protocols