Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

pHrodo™ Red Zymosan Bioparticles™ Conjugate for Phagocytosis

Catalog number:  P35364

 Related applications: Cell Viability, Proliferation & Function | Cellular Imaging | Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

New pH-sensitive pHrodo® Red dye conjugates give faster and more accurate results than any other phagocytosis assay. pHrodo® Red dye conjugates are non-fluorescent outside the cell, but fluoresce brightly red in phagosomes. Get faster staining and more accurate results—without the need for wash steps or quencher dye.

• Specific detection of phagocytosis and endocytosis
• Reduced signal variability and improved timing in sensitive experiments
• Multiplex with green dyes such as GFP, Fluo-4, or calcein

The fluorescence of the novel pHrodo® Red dye dramatically increases as pH decreases from neutral to acidic, making it an ideal tool to study phagocytosis and its regulation by drugs and/or environmental factors. The lack of fluorescence outside the cell eliminates the need for wash steps and quencher dyes.

Use the ready-made pHrodo® Red Zymosan BioParticles® Conjugate in imaging or flow applications, or pHrodo® Red SE, the activated succinimidyl ester, for labeling microorganisms or proteins of your choice.

In addition, pHrodo® dye is also available in green color, with E. coli, zymosan, S. aureus, and dextran 10,000 conjugates.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Red
Detection Method: Fluorescent
Excitation⁄Emission (nm): 560⁄585
For Use With (Equipment): Attune® Acoustic Focusing Cytometer, Confocal Microscope, Floid® Cell Imaging System, Flow Cytometer, Fluorescence Microscope, High Content Instrument
Label or Dye: pHrodo™ Red
Product Line: pHrodo™
Product Size: 5 x 1 mg
Species: S. cerevisiae
Shipping Condition: Room Temperature

Contents & storage

Store at -20 °C, dessicate and protected from light.

Documents

Manuals & protocols

Recommended products