Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Want to detect cells in hypoxic condition? Try new fixable Image-iT™ Hypoxia reagent, compatible with most fluorescent platforms.
Invitrogen™

pHrodo™ Red, succinimidyl ester (pHrodo™ Red, SE)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

New pH-sensitive pHrodo™ Red dye conjugates give faster and more accurate results than any other phagocytosis assay

pHrodo™ Red dye conjugates are non-fluorescent outside the cell, but fluoresce brightly red in phagosomes

Get faster staining and more accurate results - without the need for wash steps or quencher dye


• Specific detection of phagocytosis and endocytosis
• Reduced signal variability and improved timing in sensitive experiments
• Multiplex with green dyes such as GFP, Fluo-4, or calcein


The fluorescence of the novel pHrodo™ Red dye dramatically increases as pH decreases from neutral to the acidic, making it an ideal tool to study phagocytosis and its regulation by drugs and/or environmental factors. The lack of fluorescence outside the cell eliminates the need for wash steps and quencher dyes.

Use the ready-made pHrodo™ Red E.coli BioParticles® conjugates in imaging or flow applications, or pHrodo™ Red SE, the activated succinimidyl ester, for labeling microorganisms or proteins of your choice.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: pHrodo™ Red
Chemical Reactivity: Amine
Product Line: pHrodo™
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products