Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

ProLong™ Gold Antifade Mountant This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: P36930

Invitrogen™  Related applications: Cellular Imaging | Stem Cell Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

ProLong® Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments, and delivers this protection without significantly quenching the initial fluorescence signal. ProLong® Gold Antifade Mountant is a curing mountant that forms a near-perfect optical path (1.47 RI) and allows longer-term storage of the sample. It comes ready-to-use (room temperatures storage and no mixing required)—just apply a drop to the sample, add a coverslip, cure, and image. ProLong® Gold mountant is available with or without DAPI nuclear stain.

ProLong® Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins, such as GFP. For superior antifade protection of fluorescent proteins and fluorescent dyes, ProLong® Diamond Antifade Mountant is recommended. Learn more about key differences between ProLong® Gold and ProLong® Diamond antifade mountants.

Key Attributes:

• Protects dyes from fading during imaging
• Ready-to-use liquid that cures for longer-term storage
• Mounted samples are stable for months
• Maintains fluorescence signal—little to no quenching
• Ideal for Alexa Fluor® dyes

Get Photobleaching Protection Without Quenching
Many mountants can destroy initial fluorescence signal strength by up to 80%. To create the Molecular Probes® Gold series of mountants, our scientists developed a formula of chemical additives that minimize this initial signal quenching for the broadest range of colors and dyes.

Stable Signals During Imaging and for Longer-Term Storage
While epoxy-based mountants used in our FluoCells® series of prepared cells can be stable for years and afford the highest level of antifade protection, they are difficult to use, requiring a series of steps in organic solvents prior to mounting. ProLong® Gold Antifade Mountant is the next-best option for archiving samples. And it is the best option for preserving signal strength during prolonged imaging sessions, such as repeated scanning under confocal laser scanning illumination. It's easy to use—just air-dry your stained samples to remove excess water and mount. For immediate viewing of the sample, choose our non-curing mountant, SlowFade® Gold Antifade Mountant.

Achieve Excellent Refractive Index Matching Without Acrylic Resins
For modern high-resolution imaging applications, refractive index matching is critical. ProLong® Gold Antifade Mountant will cure to an Rf of 1.47. Additionally, the curing property means you can avoid messy acrylic resins (i.e., finger nail polish), which can damage or ruin modern microscope objectives if it comes into contact with them.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Reagent Type: Anti-Fade Solution
Product Line: ProLong®
Product Size: 10 mL
Green Features: Less hazardous, Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Storage at room temperature is recommended but can also be stored frozen (-5 to -30°C). Protect from light.

Documents

Manuals & protocols

Recommended products